3-Fluoro-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A823664-1G A823664-250MG A823664-5G A823664-100MG
77171-504EA 40 USD
77171-504 77171-506 77171-508 77893-611
3-Fluoro-2-nitrobenzaldehyde 97%
3-Fluoro-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
MDL Number: MFCD11111001
CAS Number: 872366-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR