3-Fluoro-5-iodobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-5-iodophenyl)methanol

A823363-1G A823363-250MG
77199-810EA 183.34 USD
77199-810 77199-812
3-Fluoro-5-iodobenzyl alcohol 97%
3-Fluoro-5-iodobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆FIO
MW: 252.025 g/mol
MDL Number: MFCD18396948
CAS Number: 1261837-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR