Methyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-6-carboxylate 97%

Supplier: Ambeed
Warning

A822973-100MG A822973-250MG
77199-806EA 110.21 USD
77199-806 77199-808
Methyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-6-carboxylate 97%
Methyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.06 g/mol
MDL Number: MFCD12963059
CAS Number: 1190312-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR