(R)-3-Phenyl-2-(pyrazine-2-carboxamido)propanoic acid 95%

Supplier: Ambeed
Warning

A821230-100MG A821230-250MG
77171-490EA 151.41 USD
77171-490 77171-492
(R)-3-Phenyl-2-(pyrazine-2-carboxamido)propanoic acid 95%
(R)-3-Phenyl-2-(pyrazine-2-carboxamido)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₃
MW: 271.271 g/mol
MDL Number: MFCD26960588
CAS Number: 1608986-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR