2-Chloro-1-(3-methoxyphenyl)ethanone 97%
Supplier: Ambeed
|
Danger
|
A820376-5G
A820376-1G
A820376-250MG
77199-786EA
450
USD
77199-786
77747-571
77747-572
2-Chloro-1-(3-methoxyphenyl)ethanone 97%
2-Chloro-1-(3-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₉ClO₂ |
MDL Number:
MFCD01741677 CAS Number: 82772-51-8 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...