2-Isopropyl-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A818777-100MG A818777-1G A818777-250MG A818777-5G
77199-772EA 22.66 USD
77199-772 77199-774 77199-776 77199-778
2-Isopropyl-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride 97%
2-Isopropyl-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O₂
MW: 240.68 g/mol
MDL Number: MFCD02856479
CAS Number: 1158777-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR