(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2Rp)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene

A818375-100MG A818375-50MG A818375-1G
77171-464EA 57.68 USD
77171-464 77171-466 77684-774
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene ≥98%, ee 99%
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene

Precaución: For research use only.

Formula: C₄₀H₄₄FeP₂
MW: 642.56 g/mol
MDL Number: MFCD08561161
CAS Number: 849924-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR