1-(2,5-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone 95%

Supplier: Ambeed
Warning

A818342-1G A818342-25G A818342-5G
77171-458EA 37.08 USD
77171-458 77171-460 77171-462
1-(2,5-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone 95%
1-(2,5-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₇F₂N₃O
MW: 223.18 g/mol
MDL Number: MFCD12061657
CAS Number: 1157938-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR