1-Benzyl-2-pyrrolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylpyrrolidin-2-one

A817876-100G A817876-25G A817876-5G
77199-760EA 133.9 USD
77199-760 77199-762 77199-764
1-Benzyl-2-pyrrolidinone 98%
1-Benzyl-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD00003195
CAS Number: 5291-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR