N-Methyl-(2-thienylmethyl)amine Hydrochloride 95%

Supplier: Ambeed
Warning

A817817-10G A817817-1G A817817-25G A817817-5G
77171-450EA 144.2 USD
77171-450 77171-452 77171-454 77171-456
N-Methyl-(2-thienylmethyl)amine Hydrochloride 95%
N-Methyl-(2-thienylmethyl)amine Hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClNS
MW: 163.66 g/mol
MDL Number: MFCD03306020
CAS Number: 7404-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR