2,5-Dibromo-6-fluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dibromo-2-fluoro-3-methylbenzene

A816419-100MG A816419-1G A816419-250MG
77199-744EA 7 USD
77199-744 77199-746 77199-748
2,5-Dibromo-6-fluorotoluene 98%
2,5-Dibromo-6-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
MDL Number: MFCD09800767
CAS Number: 1000576-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR