Isopropyl 2,4-dichloropyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A816136-100MG A816136-1G A816136-250MG
77171-444EA 9.27 USD
77171-444 77171-446 77171-448
Isopropyl 2,4-dichloropyrimidine-5-carboxylate 95%
Isopropyl 2,4-dichloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₂O₂
MW: 235.06 g/mol
MDL Number: MFCD28167986
CAS Number: 69312-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR