1,2-Bis(1-naphthyl)ethane 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Di(naphthalen-1-yl)ethane

A814629-100MG A814629-1G A814629-250MG A814629-5G
77199-734EA 26.78 USD
77199-734 77199-736 77199-738 77199-740
1,2-Bis(1-naphthyl)ethane 98%
1,2-Bis(1-naphthyl)ethane

Precaución: for Research Use Only

Formula: C₂₂H₁₈
MW: 282.3783 g/mol
MDL Number: MFCD00037736
CAS Number: 15374-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR