(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A813953-1G A813953-250MG A813953-5G
77171-438EA 129.78 USD
77171-438 77171-440 77171-442
(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95%
(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.08 g/mol
MDL Number: MFCD18729954
CAS Number: 1352733-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR