Benzyl Benzodithioate 98%

Supplier: Ambeed
Danger

Synonyms: Benzyl benzodithioate, Benzyl Benzenecarbodithioate, Benzodithioic acid Benzyl ester, Benzenecarbodithioic acid Benzyl ester

A813146-1G A813146-250MG A813146-25G A813146-5G
77171-430EA 155 USD
77171-430 77171-432 77171-434 77171-436
Benzyl Benzodithioate 98%
Benzyl Benzodithioate

Precaución: for Research Use Only

Formula: C₁₄H₁₂S₂
MW: 244.37 g/mol
MDL Number: MFCD08705657
CAS Number: 27249-90-7
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR