3-(Piperidin-4-yl)benzo[d]isoxazole hydrochloride 98%

Supplier: Ambeed
Warning

A812649-1G A812649-250MG A812649-5G
77171-422EA 109.18 USD
77171-422 77171-424 77171-426
3-(Piperidin-4-yl)benzo[d]isoxazole hydrochloride 98%
3-(Piperidin-4-yl)benzo[d]isoxazole hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O
MW: 238.71 g/mol
MDL Number: MFCD27978162
CAS Number: 84163-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR