(4-Bromo-1H-pyrazol-1-yl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromo-1H-pyrazol-1-yl)acetonitrile

A812251-100MG A812251-250MG A812251-5G
77199-726EA 52.53 USD
77199-726 77199-728 77747-565
(4-Bromo-1H-pyrazol-1-yl)acetonitrile 97%
(4-Bromo-1H-pyrazol-1-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₅H₄BrN₃
MW: 186.02 g/mol
MDL Number: MFCD04969698
CAS Number: 925224-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR