Methyl 3-((tert-butoxycarbonyl)amino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A811066-1G A811066-250MG A811066-5G
77199-720EA 209 USD
77199-720 77199-722 77199-724
Methyl 3-((tert-butoxycarbonyl)amino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylate 97%
Methyl 3-((tert-butoxycarbonyl)amino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₆S
MDL Number: MFCD28014041
CAS Number: 2377606-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR