2-Bromo-4-fluoro-1-nitro-3-(trifluoromethyl)benzene 97%

Supplier: Ambeed
Danger

A810547-1G A810547-5G A810547-25G
77199-712EA 108.15 USD
77199-712 77199-714 77569-514
2-Bromo-4-fluoro-1-nitro-3-(trifluoromethyl)benzene 97%
2-Bromo-4-fluoro-1-nitro-3-(trifluoromethyl)benzene

Precaución: For research use only.

Formula: C₇H₂BrF₄NO₂
MW: 287.99 g/mol
MDL Number: MFCD28740068
CAS Number: 1805937-67-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR