(6-Phenyldibenzo[b,d]furan-4-yl)boronic acid 98%

Supplier: Ambeed
Warning

A810192-1G A810192-250MG A810192-5G
77199-706EA 9 USD
77199-706 77199-708 77199-710
(6-Phenyldibenzo[b,d]furan-4-yl)boronic acid 98%
(6-Phenyldibenzo[b,d]furan-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₃BO₃
MDL Number: MFCD29077673
CAS Number: 1010068-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR