(4-Chloro-2-fluoro-3-hydroxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A810042-1G A810042-250MG A810042-5G
77199-700EA 163 USD
77199-700 77199-702 77199-704
(4-Chloro-2-fluoro-3-hydroxyphenyl)boronic acid 97%
(4-Chloro-2-fluoro-3-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₃
MDL Number: MFCD19981522
CAS Number: 1451393-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR