tert-Butyl 7-bromo-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 97%

Supplier: Ambeed
Warning

A809387-100MG A809387-1G A809387-250MG
77171-416EA 92.7 USD
77171-416 77171-418 77171-420
tert-Butyl 7-bromo-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 97%
tert-Butyl 7-bromo-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₃
MW: 314.17 g/mol
MDL Number: MFCD12913754
CAS Number: 1220039-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR