4-((4S,5R)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nutlin-3a
A809136-10MG
A809136-1MG
A809136-25MG
A809136-50MG
A809136-5MG
77199-680EA
158.62
USD
77199-680
77199-682
77199-684
77199-686
77199-688
4-((4S,5R)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one 99%
4-((4S,5R)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one
Precaución: for Research Use Only
Formula:
C₃₀H₃₀Cl₂N₄O₄ MW: 581.49 g/mol |
MDL Number:
MFCD14636430 CAS Number: 675576-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...