6-(Acetylamino)hexanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Acetamidocaproic Acid, 6-Acetamidohexanoic acid

A808730-100G A808730-100MG A808730-25G A808730-5G
77199-670EA 231.75 USD
77199-670 77199-672 77199-674 77199-676
6-(Acetylamino)hexanoic acid 98%
6-(Acetylamino)hexanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD00004424
CAS Number: 57-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR