1-[2-(4-Chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Danger

Synonyms: 4-Carboxy-1-[2-(4-chlorophenyl)ethyl]-2-oxopyrrolidine, 1-[2-(4-Carboxy-2-oxopyrrolidin-1-yl)ethyl]-4-chlorobenzene, 1-(4-Chlorophenethyl)-5-oxopyrrolidine-3-carboxylic acid

A807839-100MG A807839-1G A807839-250MG A807839-5G
77199-658EA 35.02 USD
77199-658 77199-660 77199-662 77199-664
1-[2-(4-Chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid 95%
1-[2-(4-Chlorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄ClNO₃
MW: 267.708 g/mol
MDL Number: MFCD01330893
CAS Number: 368870-06-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR