N-Cyclopropyl-1-(3-fluorophenyl)methanesulfonamide 95%
Supplier: Ambeed
|
Warning
|
A807020-100MG
A807020-1G
A807020-250MG
77171-404EA
82
USD
77171-404
77171-406
77171-408
N-Cyclopropyl-1-(3-fluorophenyl)methanesulfonamide 95%
N-Cyclopropyl-1-(3-fluorophenyl)methanesulfonamide
Precaución: for Research Use Only
Formula:
C₁₀H₁₂FNO₂S |
MDL Number:
MFCD24382615 CAS Number: 1420987-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...