(4-(Hydroxymethyl)phenyl)(phenyl)methanone 97%

Supplier: Ambeed
Warning

A806570-100MG A806570-1G
77171-400EA 49 USD
77171-400 77171-402
(4-(Hydroxymethyl)phenyl)(phenyl)methanone 97%
(4-(Hydroxymethyl)phenyl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.24 g/mol
MDL Number: MFCD26131247
CAS Number: 81449-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR