(±)-1,3-Butanediol 99%

Supplier: Ambeed
Warning

Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane

A804633-1000G A804633-100G A804633-500G
77199-638EA 25.75 USD
77199-638 77199-640 77199-642
(±)-1,3-Butanediol 99%
(±)-1,3-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
MDL Number: MFCD00004554
CAS Number: 107-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR