Ethyl 4-hydroxypicolinate 95%

Supplier: Ambeed
Warning

A804599-100MG A804599-250MG
77171-394EA 43.26 USD
77171-394 77171-396
Ethyl 4-hydroxypicolinate 95%
Ethyl 4-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD09834737
CAS Number: 53764-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR