4-Fluorothiophenol 97%

Supplier: Ambeed
Danger

Synonyms: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol

A803727-10G A803727-25G A803727-5G A803727-100G
77171-388EA 8.24 USD
77171-388 77171-390 77171-392 77684-751
4-Fluorothiophenol 97%
4-Fluorothiophenol

Precaución: For research use only.

Formula: C₆H₅FS
MW: 128.17 g/mol
MDL Number: MFCD00004846
CAS Number: 371-42-6
UN: 3336
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR