1,3-Dibromo-2,4-dichloro-5-fluorobenzene 97%

Supplier: Ambeed
Warning

A803650-100MG A803650-1G A803650-250MG A803650-5G
77199-624EA 27.81 USD
77199-624 77199-626 77199-628 77199-630
1,3-Dibromo-2,4-dichloro-5-fluorobenzene 97%
1,3-Dibromo-2,4-dichloro-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆HBr₂Cl₂F
MW: 322.78 g/mol
MDL Number: MFCD09878175
CAS Number: 1000573-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR