1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-N-methoxy-N-methylpiperidine-3-carboxamide

A803559-1G A803559-250MG A803559-5G
77199-618EA 50.47 USD
77199-618 77199-620 77199-622
1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine 97%
1-Boc-3-(N-methoxy-N-methylcarbamoyl)piperidine

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₄
MW: 272.341 g/mol
MDL Number: MFCD03069732
CAS Number: 189442-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR