1,4-Dioxane-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A802384-100MG A802384-1G A802384-250MG A802384-5G
77199-610EA 51.5 USD
77199-610 77199-612 77199-614 77199-616
1,4-Dioxane-2-carboxylic acid 95%
1,4-Dioxane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₄
MDL Number: MFCD06260725
CAS Number: 89364-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR