(Tetrahydropyran-4-yl)methyl tosylate 98%

Supplier: Ambeed
Warning

Synonyms: (Tetrahydro-2H-pyran-4-yl)methyl 4-methylbenzenesulphonate

A802146-10G A802146-1G A802146-250MG A802146-5G
77199-602EA 214.24 USD
77199-602 77199-604 77199-606 77199-608
(Tetrahydropyran-4-yl)methyl tosylate 98%
(Tetrahydropyran-4-yl)methyl tosylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₄S
MW: 270.3500 g/mol
MDL Number: MFCD09475806
CAS Number: 101691-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR