3-Acetyl-2,5-dimethylfuran 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Dimethylfuran-3-yl)ethanone

A800714-100G A800714-25G A800714-5G A800714-1G
77199-590EA 358.44 USD
77199-590 77199-592 77199-594 77747-550
3-Acetyl-2,5-dimethylfuran 98%
3-Acetyl-2,5-dimethylfuran

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00010281
CAS Number: 10599-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR