2-Fluoro-5-methoxybenzamide 97%

Supplier: Ambeed
Warning

A800695-100MG A800695-1G A800695-250MG
77199-584EA 40.17 USD
77199-584 77199-586 77199-588
2-Fluoro-5-methoxybenzamide 97%
2-Fluoro-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD06409123
CAS Number: 400-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR