10,10'-Dibromo-9,9'-bianthracene 98%

Supplier: Ambeed
Warning

A800504-1G A800504-250MG A800504-25G A800504-5G
77199-576EA 13.39 USD
77199-576 77199-578 77199-580 77199-582
10,10'-Dibromo-9,9'-bianthracene 98%
10,10'-Dibromo-9,9'-bianthracene

Precaución: for Research Use Only

Formula: C₂₈H₁₆Br₂
MW: 512.24 g/mol
MDL Number: MFCD11045035
CAS Number: 121848-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR