Dibromopyromellitic Dianhydride 98%

Supplier: Ambeed
Warning

Synonyms: 4,8-Dibromobenzo[1,2-c:4,5-c']difuran-1,3,5,7-tetraone, 3,6-Dibromobenzene-1,2,4,5-tetracarboxylic 1,2:4,5-Dianhydride

A800202-100MG A800202-1G A800202-250MG A800202-5G
77171-364EA 46 USD
77171-364 77171-366 77171-368 77171-370
Dibromopyromellitic Dianhydride 98%
Dibromopyromellitic Dianhydride

Precaución: for Research Use Only

Formula: C₁₀Br₂O₆
MW: 375.91 g/mol
MDL Number: MFCD01100205
CAS Number: 24848-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR