1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether 95%

Supplier: Ambeed
Danger

Synonyms: 1,1,2,2-Tetrafluoroethyl2,2,3,3-tetrafluoropropylether

A798265-25G A798265-5G
77199-574EA 26 USD
77199-574 77595-270
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether 95%
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether

Precaución: for Research Use Only

Formula: C₅H₄F₈O
MDL Number: MFCD00155961
CAS Number: 16627-68-2
UN: 3271
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR