3-Bromo-7-nitroquinoline 95%

Supplier: Ambeed
Warning

A797785-100MG A797785-1G A797785-250MG
77171-354EA 14 USD
77171-354 77171-356 77171-358
3-Bromo-7-nitroquinoline 95%
3-Bromo-7-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O₂
MDL Number: MFCD28133455
CAS Number: 1354221-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR