3,8-Dibromo-1,10-phenanthroline-5,6-dione 97%

Supplier: Ambeed
Warning

A797586-100MG A797586-1G A797586-250MG A797586-5G
77199-566EA 20.6 USD
77199-566 77199-568 77199-570 77199-572
3,8-Dibromo-1,10-phenanthroline-5,6-dione 97%
3,8-Dibromo-1,10-phenanthroline-5,6-dione

Precaución: for Research Use Only

Formula: C₁₂H₄Br₂N₂O₂
MW: 367.98 g/mol
MDL Number: MFCD09953785
CAS Number: 602331-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR