1,10-Phenanthroline-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A795596-100MG A795596-1G A795596-250MG
77199-558EA 45 USD
77199-558 77199-560 77199-562
1,10-Phenanthroline-5-carboxylic acid 95%
1,10-Phenanthroline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈N₂O₂
MDL Number: MFCD06255104
CAS Number: 630067-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR