4-(3-bromophenethyl)morpholine 95%

Supplier: Ambeed
Warning

A792920-100MG A792920-1G A792920-250MG
77171-322EA 41.2 USD
77171-322 77171-324 77171-326
4-(3-bromophenethyl)morpholine 95%
4-(3-bromophenethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO
MW: 270.17 g/mol
MDL Number: MFCD25962760
CAS Number: 364793-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR