3-Hydroxy-N,N-dimethylpicolinamide 95%

Supplier: Ambeed
Warning

A790316-100MG A790316-1G A790316-250MG
77171-314EA 16 USD
77171-314 77171-316 77171-318
3-Hydroxy-N,N-dimethylpicolinamide 95%
3-Hydroxy-N,N-dimethylpicolinamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.17 g/mol
MDL Number: MFCD12808037
CAS Number: 1076-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR