1-Methylimidazolium tetrafluoroborate 95%

Supplier: Ambeed
Danger

A790171-1G A790171-25G A790171-5G
77171-308EA 10 USD
77171-308 77171-310 77171-312
1-Methylimidazolium tetrafluoroborate 95%
1-Methylimidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₄H₇BF₄N₂
MDL Number: MFCD09038841
CAS Number: 151200-14-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR