2-Methyl-2,6-diazaspiro[3.3]heptane dihydrochloride 96%

Supplier: Ambeed
Warning

A789398-100MG A789398-1G A789398-250MG A789398-5G
77171-302EA 101.97 USD
77171-302 77171-304 77171-306 77684-732
2-Methyl-2,6-diazaspiro[3.3]heptane dihydrochloride 96%
2-Methyl-2,6-diazaspiro[3.3]heptane dihydrochloride

Precaución: For research use only.

Formula: C₆H₁₄Cl₂N₂
MW: 185.09 g/mol
MDL Number: MFCD26954754
CAS Number: 1630082-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR