Ethyl-11-bromoundecanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 11-bromoundecanoate, 11-Bromoundecanoic Acid Ethyl Ester

A789193-1G A789193-25G A789193-5G
77199-522EA 32.96 USD
77199-522 77199-524 77199-526
Ethyl-11-bromoundecanoate 98%
Ethyl-11-bromoundecanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₅BrO₂
MW: 293.25 g/mol
MDL Number: MFCD00045048
CAS Number: 6271-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR