1,4,7,10-Tetraazacyclododecane tetrahydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Cyclen tetrahydrochloride

A789089-100G A789089-25G A789089-5G
77199-514EA 156.56 USD
77199-514 77199-516 77199-518
1,4,7,10-Tetraazacyclododecane tetrahydrochloride 98%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₄Cl₄N₄
MDL Number: MFCD00068484
CAS Number: 10045-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR