1,4-Dimethoxy-2-butyne 98%

Supplier: Ambeed
Danger

A788848-100MG A788848-1G A788848-250MG A788848-5G
77199-506EA 6 USD
77199-506 77199-508 77199-510 77199-512
1,4-Dimethoxy-2-butyne 98%
1,4-Dimethoxy-2-butyne

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MDL Number: MFCD00048069
CAS Number: 16356-02-8
UN: 3271
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR