Tetrafluorophthalic anhydride 98%

Supplier: Ambeed
Warning

A788636-100G A788636-25G A788636-5G
77199-494EA 346 USD
77199-494 77199-496 77199-498
Tetrafluorophthalic anhydride 98%
Tetrafluorophthalic anhydride

Precaución: for Research Use Only

Formula: C₈F₄O₃
MDL Number: MFCD00039697
CAS Number: 652-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR